ENA295068412 Enamine

3-(1,2,3,4-tetrahydroisoquinoline-2-carbonyl)phenol

  • MDL number MFCD12621750
  • Cas Number : 1019438-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63866
Availability details for
Quantity
  • Availability
  • Add to Cart