ENA295068415 Enamine

3-(1,2-dimethyl-1H-imidazole-4-sulfonamido)propanoic acid

  • MDL number MFCD16547514
  • Cas Number : 1240526-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58950
Availability details for
Quantity
  • Availability
  • Add to Cart