ENA295068417 Enamine

2-(1-methyl-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD09925217
  • Cas Number : 1152582-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63872

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-63872
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart