ENA295068420 Enamine

[1-(5-methylfuran-2-carbonyl)piperidin-3-yl]methanol

  • MDL number MFCD12143786
  • Cas Number : 1156314-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63876
Availability details for
Quantity
  • Availability
  • Add to Cart