ENA295068424 Enamine

2-amino-N-[3-(pyrrolidin-1-yl)phenyl]pentanamide

  • MDL number MFCD12834234
  • Cas Number : 1218021-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63882
Availability details for
Quantity
  • Availability
  • Add to Cart