ENA295068427 Enamine

1-cyclopropyl-1-[4-(2-methylpropyl)phenyl]methanamine

  • MDL number MFCD05191988
  • Cas Number : 902742-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63885
Availability details for
Quantity
  • Availability
  • Add to Cart