ENA295068437 Enamine

6-(3-methoxypropoxy)pyridine-3-carboxylic acid

  • MDL number MFCD12767860
  • Cas Number : 1183915-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63907
Availability details for
Quantity
  • Availability
  • Add to Cart