ENA295068438 Enamine

4-(4-fluorophenyl)-1,3-thiazol-5-amine

  • MDL number MFCD16547591
  • Cas Number : 1240529-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63908

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63908
Availability details for
Quantity
  • Availability
  • Add to Cart