ENA295068441 Enamine

N-(4-cyanophenyl)piperidine-4-carboxamide

  • MDL number MFCD09815017
  • Cas Number : 1016734-18-1
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-cyanophenyl)-4-piperidinecarboxamide
Inchi Code 1S/C13H15N3O/c14-9-10-1-3-12(4-2-10)16-13(17)11-5-7-15-8-6-11/h1-4,11,15H,5-8H2,(H,16,17)
Inchi Key NUPOLOKBKIZZPM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63912
Availability details for
Quantity
  • Availability
  • Add to Cart