ENA295068444 Enamine

3-(3-methyl-1H-pyrazol-1-yl)aniline

  • MDL number MFCD09814329
  • Cas Number : 1134414-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63915
Availability details for
Quantity
  • Availability
  • Add to Cart