ENA295068450 Enamine

2-(3,4-dichlorophenyl)-2-(methylamino)acetic acid

  • MDL number MFCD12047908
  • Cas Number : 1218475-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63922
Availability details for
Quantity
  • Availability
  • Add to Cart