ENA295068460 Enamine

methyl 3-amino-5-fluoro-4-methylbenzoate

  • MDL number MFCD14694719
  • Cas Number : 1093087-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60390
Availability details for
Quantity
  • Availability
  • Add to Cart