ENA295068464 Enamine

5,7-diazaspiro[3.4]octane-6,8-dione

  • MDL number MFCD09045753
  • Cas Number : 89691-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-225
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-63937

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P305+P351+P338,P330,P332+P313,P337+P313,P362,P501
hcodes Xi,Xi
MSDS https://www.enaminestore.com/catalog/EN300-63937
pictograms GHS07
hstatements H302,H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart