ENA295068466 Enamine

5-[2-(2H-1,3-benzodioxol-5-yl)ethyl]-5-methylimidazolidine-2,4-dione

  • MDL number MFCD09042563
  • Cas Number : 1052562-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63940

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63940
Availability details for
Quantity
  • Availability
  • Add to Cart