ENA295068474 Enamine

2-(thiophen-2-yl)imidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD15732273
  • Cas Number : 1105195-06-9
  • Molecular Weight : 215.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63954
Availability details for
Quantity
  • Availability
  • Add to Cart