ENA295068476 Enamine

4-(3-chloropropyl)benzene-1-sulfonyl chloride

  • MDL number MFCD10034229
  • Cas Number : 7296-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-63961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63961
Availability details for
Quantity
  • Availability
  • Add to Cart