ENA295068479 Enamine

5-(3,5-dichlorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD16547607
  • Cas Number : 766520-00-7
  • Molecular Weight : 228.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63970
Availability details for
Quantity
  • Availability
  • Add to Cart