ENA295068484 Enamine

N-(3-carbamothioylphenyl)acetamide

  • MDL number MFCD09939039
  • Cas Number : 93029-92-6
  • Molecular Weight : 194.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63976
Availability details for
Quantity
  • Availability
  • Add to Cart