ENA295068485 Enamine

3-methyl-5-(3-methylphenyl)-1H-1,2,4-triazole

  • MDL number MFCD11215138
  • Cas Number : 1094688-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63977
Availability details for
Quantity
  • Availability
  • Add to Cart