ENA295068488 Enamine

N-methyl-2-(methylamino)-N-(pyridin-2-yl)acetamide

  • MDL number MFCD11128717
  • Cas Number : 1019325-08-6
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63980
Availability details for
Quantity
  • Availability
  • Add to Cart