ENA295068493 Enamine

2-methylpropyl 4-(2-aminophenyl)piperazine-1-carboxylate

  • MDL number MFCD12670836
  • Cas Number : 136817-34-0
  • Molecular Weight : 277.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63989
Availability details for
Quantity
  • Availability
  • Add to Cart