ENA295068495 Enamine

2-(3-amino-1H-1,2,4-triazol-1-yl)-N-benzylacetamide

  • MDL number MFCD12901416
  • Cas Number : 1179649-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63993
Availability details for
Quantity
  • Availability
  • Add to Cart