ENA295068497 Enamine

2,5-dimethyl 3-hydroxythiophene-2,5-dicarboxylate

  • MDL number MFCD04110180
  • Cas Number : 5556-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63996
Availability details for
Quantity
  • Availability
  • Add to Cart