ENA295068499 Enamine

2-[3-(difluoromethoxy)-4-methoxyphenyl]ethan-1-amine

  • MDL number MFCD09814408
  • Cas Number : 823234-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64000
Availability details for
Quantity
  • Availability
  • Add to Cart