ENA295068501 Enamine

2-[2-(4-methylphenyl)-1,3-thiazol-4-yl]ethan-1-amine

  • MDL number MFCD09724090
  • Cas Number : 871689-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64002
Availability details for
Quantity
  • Availability
  • Add to Cart