ENA295068503 Enamine

2-(benzylamino)benzonitrile

  • MDL number MFCD00512083
  • Cas Number : 5589-62-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32913
Availability details for
Quantity
  • Availability
  • Add to Cart