ENA295068507 Enamine

4-[(dimethylamino)methyl]benzoic acid hydrochloride

  • MDL number MFCD09997604
  • Cas Number : 17847-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64034

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64034
Availability details for
Quantity
  • Availability
  • Add to Cart