ENA295068508 Enamine

2-(2,6-dichlorophenyl)propanoic acid

  • MDL number MFCD11036862
  • Cas Number : 2012-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64035
Availability details for
Quantity
  • Availability
  • Add to Cart