ENA295068510 Enamine

N-ethyl-2-(4-formylphenoxy)acetamide

  • MDL number MFCD08444984
  • Cas Number : 182686-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64037
Availability details for
Quantity
  • Availability
  • Add to Cart