ENA295068513 Enamine

[2-(pyridin-2-yl)ethyl]urea

  • MDL number MFCD09944932
  • Cas Number : 20273-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64041
Availability details for
Quantity
  • Availability
  • Add to Cart