ENA295068515 Enamine

3-{[(1-cyclopropylethyl)amino]methyl}phenol

  • MDL number MFCD12193533
  • Cas Number : 1154734-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64045
Availability details for
Quantity
  • Availability
  • Add to Cart