ENA295068519 Enamine

[5-(4-fluorophenyl)-1,3-oxazol-2-yl]methanol

  • MDL number MFCD12137937
  • Cas Number : 844818-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64051
Availability details for
Quantity
  • Availability
  • Add to Cart