ENA295068520 Enamine

5-(5-methylthiophen-2-yl)-1,2,4-triazin-3-amine

  • MDL number MFCD11134123
  • Cas Number : 1094454-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64052
Availability details for
Quantity
  • Availability
  • Add to Cart