ENA295068523 Enamine

4,5-dimethoxy-2-methylaniline

  • MDL number MFCD00130067
  • Cas Number : 41864-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-34110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34110
Availability details for
Quantity
  • Availability
  • Add to Cart