ENA295068529 Enamine

1-cyclohexylpropan-1-one

  • MDL number MFCD00060814
  • Cas Number : 1123-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64061
Availability details for
Quantity
  • Availability
  • Add to Cart