ENA295068537 Enamine

1,2-dimethyl-4-phenyl-1H-imidazol-5-amine

  • MDL number MFCD11186827
  • Cas Number : 1040040-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61342

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61342
Availability details for
Quantity
  • Availability
  • Add to Cart