ENA295068539 Enamine

6-(4-methyl-1,4-diazepan-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD11169412
  • Cas Number : 910108-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61395
Availability details for
Quantity
  • Availability
  • Add to Cart