ENA295068544 Enamine

1-[(3,5-dimethyl-1H-pyrazol-4-yl)sulfonyl]-4-methylpiperidine

  • MDL number MFCD07366377
  • Cas Number : 1023947-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35312
Availability details for
Quantity
  • Availability
  • Add to Cart