ENA295068546 Enamine

3-(4-fluorophenyl)-2-oxopropanoic acid

  • MDL number MFCD09732983
  • Cas Number : 7761-30-0
  • Molecular Weight : 182.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64097
Availability details for
Quantity
  • Availability
  • Add to Cart