ENA295068550 Enamine

1-(2-aminoethyl)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD09805470
  • Cas Number : 933739-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64101

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64101
Availability details for
Quantity
  • Availability
  • Add to Cart