ENA295068552 Enamine

N'-hydroxynaphthalene-1-carboximidamide

  • Cas Number : 40019-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61644
Availability details for
Quantity
  • Availability
  • Add to Cart