ENA295068553 Enamine

2-amino-1-cyclohexylethan-1-ol

  • MDL number MFCD13687632
  • Cas Number : 57230-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36150
Availability details for
Quantity
  • Availability
  • Add to Cart