ENA295068554 Enamine

tert-butyl 4-[(4-bromophenyl)amino]piperidine-1-carboxylate

  • MDL number MFCD07785997
  • Cas Number : 443998-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64107
Availability details for
Quantity
  • Availability
  • Add to Cart