ENA295068556 Enamine

3-(1-hydroxyethyl)benzene-1-sulfonamide

  • MDL number MFCD12186255
  • Cas Number : 210827-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61679
Availability details for
Quantity
  • Availability
  • Add to Cart