ENA295068557 Enamine

(1-phenyl-1H-1,2,3-triazol-4-yl)methanol

  • MDL number MFCD00100214
  • Cas Number : 103755-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64110
Availability details for
Quantity
  • Availability
  • Add to Cart