ENA295068562 Enamine

1-(4-bromo-2-fluorophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD09934502
  • Cas Number : 530080-21-8
  • Molecular Weight : 243.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61813
Availability details for
Quantity
  • Availability
  • Add to Cart