ENA295068564 Enamine

[2-(4-methoxyphenyl)ethyl]urea

  • MDL number MFCD01675975
  • Cas Number : 13576-85-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64117
Availability details for
Quantity
  • Availability
  • Add to Cart