ENA295068565 Enamine

5-(3-bromophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD10006415
  • Cas Number : 1017473-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64119
Availability details for
Quantity
  • Availability
  • Add to Cart