ENA295068569 Enamine

2-[(1,3-thiazol-4-yl)methoxy]phenol

  • MDL number MFCD11181813
  • Cas Number : 1094247-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64123
Availability details for
Quantity
  • Availability
  • Add to Cart