ENA295068575 Enamine

1-(2,6-dimethylpyrimidin-4-yl)-N-methylpiperidin-4-amine

  • MDL number MFCD12768724
  • Cas Number : 1184506-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61848

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61848
Availability details for
Quantity
  • Availability
  • Add to Cart